Learn More
Mirus Bio™ Ingenio™ Electroporation Solution
Proprietary Ingenio™ Electroporation Solution facilitates efficient and reliable delivery of nucleic acids to eukaryotic cells resistant to chemical transfection.
Supplier:  Mirus Bio™ MIR 50114
 View more versions of this product
Catalog No. MIR50114
Only null left
Description
Description
  • Broad spectrum solution supports high efficiency electroporation of hard to transfect cells with minimal toxicity
  • Compatible with all electroporation devices: Use with your existing system; no need to purchase additional specialized equipment
  • Replaces standard electroporation solutions including phosphate buffered saline and serum-free media
  • High cell viability: Minimize the risk of introducing experimental bias due to toxicity-induced cellular changes
Specifications
Chemical Identifiers
Electroporation
Electroporation Solution
4°C
All electroporation device
Sterile
Exponential Decay Wave
Specifications
Electroporation
Electroporation Solution
12.5 mL
4°C
All electroporation device
50 Reactions
Sterile
Exponential Decay Wave
Product Suggestions
Product Suggestions