missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Inc Influenza B (Austria/1359417/2021) Neuraminidase (NA) Protein, His Tag, 100 g

Catalog No. NC2387742
Encompass

Recombinant neuraminidase (NA) from Influenza B (Austria/1359417/2021), expressed in HEK293 cells with an N-terminal His tag. The glycoprotein cleaves sialic acid residues to facilitate viral release and is suitable for bioactivity, binding, and assay development. Purity and molecular characteristics have been verified by SDS-PAGE and SEC-MALS.

  • Available in 100 g and 1 mg sizes.
  • Expressed in human 293 (HEK293) cells.
  • N-terminal polyhistidine (His) tag.
  • Purity >90% by SDS-PAGE; SEC-MALS verification.
  • Molecular weight ~260-290 kDa (SEC-MALS verified).
  • Validated in bioactivity assays including SPR binding to Neu5Ac.

Catalog No. NC2387742 Supplier ACROBiosystems Inc Supplier No. NEEV5245100UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Recombinant neuraminidase (NA) from Influenza B (Austria/1359417/2021), expressed in HEK293 cells with an N-terminal His tag. The glycoprotein cleaves sialic acid residues to facilitate viral release and is suitable for bioactivity, binding, and assay development. Purity and molecular characteristics have been verified by SDS-PAGE and SEC-MALS.

  • Available in 100 g and 1 mg sizes.
  • Expressed in human 293 (HEK293) cells.
  • N-terminal polyhistidine (His) tag.
  • Purity >90% by SDS-PAGE; SEC-MALS verification.
  • Molecular weight ~260-290 kDa (SEC-MALS verified).
  • Validated in bioactivity assays including SPR binding to Neu5Ac.