missing translation for 'onlineSavingsMsg'
Learn More

SOMNI Scientific™ InductFlow-LA Large RED Active Vacuum Style Induction Chamber

Catalog No. 04777120
Change view
Click to view available options
For Use With (Application):
Research Animal Inhalant Anesthesia
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
04777120 Research Animal Inhalant Anesthesia
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04777120 Supplier SOMNI Scientific™ Supplier No. IR3010

The red-tinted shatter-resistant acrylic allows for subject observation while reducing stress during anesthetic exposure. Must be used with attenuated active waste gas removal.

  • Active style prevents unnecessary exposure when used with active waste gas removal system
  • Red tint creates less stressful environment
  • Slide top lid
  • One-handed operation
  • Color coded inlet and outlet ports
  • Compatible with 19 mm tubing & 15 mm adapters

Specifications

For Use With (Application) Research Animal Inhalant Anesthesia
For Use With (Equipment) AMD-17106, TT-17000, TT-17001, TT-17002, AMD-17004, WG-15001, WG-15101
Material 1/4 in. Acrylic
Length (Metric) 29.85 cm
Width (Metric) 15.24 cm
Includes Color-coded ports

For Veterinary Use Only