missing translation for 'onlineSavingsMsg'
Learn More

Indium tin oxide, 99.99% (metals basis)

Catalog No. AA3634822
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3634822 100 g
AA3634806 5 g
AA3634814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3634822 Supplier Thermo Scientific Chemicals Supplier No. 03634822
Only null left

CAS: 50926-11-9 | InOSn | 249.527 g/mol

Indium tin oxide is used in nanotechnology to provide a path to a new generation of solar cells, to make transparent conductive coatings for displays such as liquid crystal displays, flat panel displays, plasma displays, touch panels, and electronic ink. Thin films of ITO are also used in organic light-emitting diodes, antistatic coatings and EMI shielding. In organic light-emitting diodes, ITO is used as the anode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50926-11-9
Molecular Formula InOSn
Molecular Weight (g/mol) 249.527
MDL Number MFCD00171662
InChI Key AMGQUBHHOARCQH-UHFFFAOYSA-N
Synonym acmc-1au9m, tin ii oxide indigane
PubChem CID 57350325
IUPAC Name indium;oxotin
SMILES O=[Sn].[In]

Specifications

Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Insoluble in water.
Odor Odorless
Appearance White powder
Chemical Name or Material Indium tin oxide
Hazard Category H335-H351
Precautionary Statement P201-P202-P261-P271-P281-P304+P340-P308+P313-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only