missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) chloride, anhydrous, 99.999% (metals basis)

Catalog No. AA1185630
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1185630 250 g
AA1185603 1 g
AA1185609 10 g
AA1185618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA1185630 Supplier Thermo Scientific Chemicals Supplier No. 01185630
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 10025-82-8

It is used in famous organic reactions such as Friedel-Crafts acylation and Diels-Alder reaction as a lewis acid catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10025-82-8
MDL Number MFCD00011058

Specifications

Boiling Point 800°C
Melting Point 586°C
Color White
Physical Form Crystalline
Quantity 250 g
UN Number UN3260
Merck Index 14,4958
Solubility Information It is very soluble in water and THF. It is sparingly soluble in alcohol and insoluble in ether.
Formula Weight 221.18
Percent Purity 99.999%
Odor Odorless
Sensitivity Hygroscopic
Density 3.460 g/mL
Assay (metals basis)
Chemical Name or Material Indium(III) chloride, Anhydrous
Show More Show Less
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 233-043-0
RTECSNumber NL1400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only