missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) acetate, 99.99% (metals basis)

Catalog No. AA4223018
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4223018 50 g
AA4223009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4223018 Supplier Thermo Scientific Chemicals Supplier No. 04223018

CAS: 25114-58-3 | C2H4InO2 | 174.87 g/mol

Indium(III) acetate is used as a catalyst in intermolecular radical addition of organic iodides to electron deficient alkenes. It is also used in the reduction of 1-bromo-3-phenylpropane to propylbenzene. Further, it is used to prepare the indium oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25114-58-3
Molecular Formula C2H4InO2
Molecular Weight (g/mol) 174.87
MDL Number MFCD00054347
InChI Key LGHOYKSQIQISBI-UHFFFAOYSA-N
Synonym indium acetate, acetic acid, indium 3+ salt
PubChem CID 24191761
IUPAC Name acetic acid;indium
SMILES CC(=O)O.[In]

Specifications

Physical Form Powder
Quantity 50 g
Assay Percent Range 99.99% (Metals Basis)
Solubility Information Soluble in water,mineral acid and acetic acid.
Formula Weight 291.96
Sensitivity Hygroscopic
Chemical Name or Material Indium(III) Acetate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only