missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indirubin 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 350°C |
| Color | Purple-Red |
| Quantity | 25 mg |
| Formula Weight | 262.27 |
| Percent Purity | ≥97.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Indirubin |
Safety and Handling
| RTECSNumber | DU2995000 |
| TSCA | No |
| Recommended Storage | Refrigerator |