missing translation for 'onlineSavingsMsg'
Learn More

Indirubin-3'-oxime, 98%, Thermo Scientific Chemicals

Catalog No. AAJ67459LB0
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67459LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ67459LB0 Supplier Thermo Scientific Chemicals Supplier No. J67459LB0

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 160807-49-8
Molecular Formula C16H11N3O2
Molecular Weight (g/mol) 277.28
MDL Number MFCD02683594
InChI Key LDEWQRSYYHTQRA-UHFFFAOYSA-N
Synonym indirubin-3'-monoxime, indirubin-3'-oxime, indirubin-3-oxime, indirubin-3monoxime, indirubin-3'-monooxime, indirubin-3-monoxime, 3-3-hydroxyamino-1h-indol-2-yl indol-2-one, indirubin-3′-monoxime, z-1h,1'h-2,3' biindolylidene-3,2'-dione-3-oxime, 3-1,3-dihydro-3-hydroxyimino-2h-indol-2-ylidene-1,3-dihydro-2h-indol-2-one
PubChem CID 5326739
IUPAC Name 3-(hydroxyamino)-1H,2'H-[2,3'-biindol]-2'-one
SMILES ONC1=C(NC2=CC=CC=C12)C1=C2C=CC=CC2=NC1=O

Specifications

Color Red
Quantity 5 mg
Formula Weight 277.28
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Indirubin-3'-oxime
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only