missing translation for 'onlineSavingsMsg'
Learn More

Indigodisulfonic Acid Dipotassium Salt 90.0+%, TCI America™

Catalog No. I02231G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I02231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I02231G Supplier TCI America Supplier No. I02231G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13725-33-2
Molecular Formula C16H8K2N2O8S2
Molecular Weight (g/mol) 498.563
MDL Number MFCD00067311
InChI Key PLXHLMUZUKUFAR-QDBORUFSSA-L
PubChem CID 25113449
IUPAC Name dipotassium;(2E)-3-oxo-2-(3-oxo-5-sulfonato-1H-indol-2-ylidene)-1H-indole-5-sulfonate
SMILES C1=CC2=C(C=C1S(=O)(=O)[O-])C(=O)C(=C3C(=O)C4=C(N3)C=CC(=C4)S(=O)(=O)[O-])N2.[K+].[K+]

Specifications

Color Blue-Purple
Quantity 1 g
Formula Weight 498.56
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Indigodisulfonic Acid Dipotassium Salt
EINECSNumber (5)-1650
TSCA No