missing translation for 'onlineSavingsMsg'
Learn More

Indene 93.0+%, TCI America™

Catalog No. I0016500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0016500ML 500 mL
I001625ML 25 mL
I0016100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I0016500ML Supplier TCI America Supplier No. I0016500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 95-13-6
Molecular Formula C9H8
Molecular Weight (g/mol) 116.163
MDL Number MFCD00003777
InChI Key YBYIRNPNPLQARY-UHFFFAOYSA-N
Synonym indene, indonaphthene, inden, unii-67h8y6lb8a, indenyl radical, 1h-inden, pubchem20728, indene, reag, acmc-209ruy
PubChem CID 7219
ChEBI CHEBI:41921
IUPAC Name 1H-indene
SMILES C1C=CC2=CC=CC=C21

Specifications

Color Yellow
Boiling Point 180°C
Quantity 500 mL
UN Number 3295
Formula Weight 116.16
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material Indene
EINECSNumber (4)-0580
RTECSNumber NK8225000
TSCA Yes
Recommended Storage Freezer