missing translation for 'onlineSavingsMsg'
Learn More

Indapamide, MilliporeSigma™ Supelco™

Indapamide is a deuretic drug, belonging to the class of benzothiadiazines. It is commonly used for the treatment of hypertension. The drug acts by causing a drop in systolic, diastolic and mean blood pressure.

$118.97 - $550.43

Specifications

CAS 26807-65-8
Physical Form Neat
Linear Formula C16H16 ClN3O3 S
MDL Number MFCD00079375
Synonym N-(4-Chloro-3-sulfamoylbenzamido)-2-methylindoline
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111013783
View Documents
Merck Emd Millipore
I1887250MG
250 mg
Each for $118.97
Only null left
 
111013782
View Documents
Merck Emd Millipore
I18875G
5 g
Each for $550.43
Only null left
 
Description

Description

Featured Industries: Pharmaceutical (small molecule)
Specifications

Specifications

26807-65-8
C16H16 ClN3O3 S
N-(4-Chloro-3-sulfamoylbenzamido)-2-methylindoline
Drug Analysis
Analytical Standard
Neat
MFCD00079375
Analytical Standard
365.83
Videos
SDS
Documents

Documents