missing translation for 'onlineSavingsMsg'
Learn More

Imperatorin 98.0+%, TCI America™

Catalog No. I090425MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I090425MG 25 mg
I0904100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I090425MG Supplier TCI America Supplier No. I090425MG
Only null left

Chemical Identifiers

CAS 482-44-0
Molecular Formula C16H14O4
Molecular Weight (g/mol) 270.284
MDL Number MFCD00016881
InChI Key OLOOJGVNMBJLLR-UHFFFAOYSA-N
Synonym 8-(3-Methyl-2-butenyloxy)psoralen, 8-Prenyloxypsoralen, Marmelosin, 9-[(3-Methyl-2-buten-1-yl)oxy]-7H-furo[3,2-g]chromen-7-one
PubChem CID 10212
ChEBI CHEBI:5885
IUPAC Name 9-(3-methylbut-2-enoxy)furo[3,2-g]chromen-7-one
SMILES CC(=CCOC1=C2C(=CC3=C1OC=C3)C=CC(=O)O2)C

Specifications

Melting Point 99°C
Quantity 25 mg
Formula Weight 270.28
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Imperatorin
RTECSNumber LV1575000
TSCA No
Recommended Storage Freezer