Learn More
Alkali Scientific ASI™ Immunoplate (ELISA) Strip Single Well Plate, PS, Flat Bottom, 8 Well, 100/Cs, Medibinding
Numéro de catalogue. 503028314
Encompass
Multibinding :Suitable specifically for adsorbing hydrophilic molecules. (Antigen ELISA) Maxibinding :Modified polystyrene surface for higher binding capacity for proteins and other molecules with both hydrophilic and hydrophobic regions. (Recommended for most ELISA experiments) Medibinding :Surface adsorbs proteins and molecules that consists of interrmediate hydrophobic and hydrophilic properties.(Antigen/Antibody ELISA; More hydrophbic compared to Maxibinding Plates) Unibinding :Suitable for adsorption of hydrophobic molecules. (Antigen ELISA) Strip single well type (8well/strip, breakable well)Lid not included Developed for immunoassay(ELISA) and general binding assay Uniform surface homogeneity Flat bottom Raised rims on the wells Alphanumeric labelling Designed for easy handling and automation
Numéro de catalogue. 503028314 Fournisseur Alkali Scientific Code fournisseur SSWMEDI
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Multibinding :Suitable specifically for adsorbing hydrophilic molecules. (Antigen ELISA) Maxibinding :Modified polystyrene surface for higher binding capacity for proteins and other molecules with both hydrophilic and hydrophobic regions. (Recommended for most ELISA experiments) Medibinding :Surface adsorbs proteins and molecules that consists of interrmediate hydrophobic and hydrophilic properties.(Antigen/Antibody ELISA; More hydrophbic compared to Maxibinding Plates) Unibinding :Suitable for adsorption of hydrophobic molecules. (Antigen ELISA) Strip single well type (8well/strip, breakable well)Lid not included Developed for immunoassay(ELISA) and general binding assay Uniform surface homogeneity Flat bottom Raised rims on the wells Alphanumeric labelling Designed for easy handling and automation