missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Immuno Clear Standard Modules, Max. working volume: 330μL
GREENER_CHOICE

Catalog No. 14245163
Change view
Click to view available options
Surface Treatment:
Immulon 4 HBX
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Surface Treatment
14245163 Immulon 4 HBX
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14245163 Supplier Thermo Scientific™ Supplier No. 6404

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Immuno Clear Standard Modules feature Nunc™, Immulon™ and Microtiter™ module plates that are developed for ELISA techniques and fit standard equipment.

Thermo Scientific™ Immuno Clear Standard Modules feature Nunc™, Immulon™ and Microtiter™ module plates that are developed for ELISA techniques and fit standard equipment. Their format provides flexible use of components with small sample sizes. The modules offer lot-to-lot reliability, well-to-well reproducibility and are backed by industry-leading quality standards and certifications.

Thermo Scientific standard modules are strip plates with solid modules that can be easily inserted and removed from their frame. This enhances the use of the plate with smaller sample sizes.

  • Several strip formats: 8-well strip, 12-well strip, 16-well strip (two columns of 8 wells)
  • Several well shapes: flat bottom, C bottom (radius edge), U shape wells, StarWells (wells have fins to increase the surface area and enhance performance)
  • Several surfaces to maximize assay performance
  • Modules remain in frame when inverted
  • Stackable frame marked for lid orientation
  • Polystyrene wells in polyethylene frame
  • Ext. Dim.: 128 × 86mm

Choice of Surface

  • PolySorp™, Immulon 1B, and Universal Binding (UB) for adsorption of hydrophobic molecules
  • Medisorp™ and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules
  • MaxiSorp™, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays
  • MultiSorp™ are very hydrophilic and works well for glycans and water soluble proteins

Ordering Information:

Free-standing modules. Cat. No. 95029440 and 464394 are certified. Frames and modules available separately. Sealing tape and strip caps are available. Color-coded modules are available on request. Contact your Customer Service or Sales Representative for further details.

Specifications

Breakapart Breakable Strips
Binding Property Hydrophilic
Certifications/Compliance Certified for adsorption of IgG; CV of <5% between wells
Certified Yes
Detection Method Colorimetric
Packaging 320 Pk., 320 Cases
Binding Type Adsorption
Color Clear
Product Type Unframed – Use Frame 6604
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 330 μL
Surface Treatment Immulon 4 HBX
Well Shape Round, Flat Bottom (F-well)
No. per Case 320
No. per Pack 320
Plate Blocking None
Target Molecule Antibodies (Glycoproteins)
Well Design F12
Show More Show Less