ImmPrint Permanent Marking Pen, The high density, Fast-drying black ink is resistant to most organic solvents encountered in histological applications, such as alcohols and clearing agents, 5/ST
Numéro de catalogue.
NC9279940
Fournisseur
Vector LaboratoriesCode fournisseur
H6100
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
ImmPrint Permanent Marking Pen, The high density, Fast-drying black ink is resistant to most organic solvents encountered in histological applications, such as alcohols and clearing agents, 5/ST