missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Immobilon-NC Membranes, Triton-Free, 0.45 μm

Mixed Cellulose Esters, Surfactant-free

Supplier:  MilliporeSigma™ HATF08250

 View more versions of this product

Catalog No. HATF08250


Only null left

Description

Description

Mixed Cellulose Esters, Surfactant-free

0.45 μm (0.45 micron filter) pore size Mixed Cellulose Esters, Surfactant-Free, Transfer Membrane for western blotting using Chromogenic, Radioactive or Fluorescent detection methods.
Specifications

Specifications

Not Autoclavable
MCE
Non-sterile
Clarification of fluids; Sterilizing filtration of tissue culture media, additives, antibiotics; Air monitoring; Particulate analysis; Particulate removal; Biological assays (receptor binding studies, TCA precipitates, etc.).
Blotting Membranes
82 mm
White
0.45 μm
60 mL/min x cm2
Plain
Hydrophilic
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions