missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Immobilon-E Blotting Sandwich

MilliporeSigma™ Immobilion™-E Blotting Sandwiches are a convenient way to process multiple Western Blots

Specifications

Description Easily wetted with water
For Use With (Application) Western Blotting
Color White
Code IEVH
Surface Plain
View More Specs

Products 2
Catalog Number Mfr. No. Dimensions (LxW) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (LxW) Price Quantity & Availability  
IESN07852
View Documents Promotion Details
MilliporeSigma™
IESN07852
7 x 8.4 cm N/A
Only null left
 
IESN08132
View Documents Promotion Details
MilliporeSigma™
IESN08132
8.5 x 13.5 cm N/A
Only null left
 
Description

Description

Features:

  • 0.45 μm pore size is ideal for most Western blotting applications with chemiluminescence or chromogenic detection
  • Saves time and reduces waste by eliminating the alcohol pre-wet step
  • Higher protein retention delivers stronger signal than most nitrocellulose membranes
  • Sandwiches include sheets of Immobilon™-E PVDF transfer membrane interleaved with pre-cut sheets of chromatography-grade blotting filter paper. 
Specifications

Specifications

Easily wetted with water
White
Plain
PVDF
Western Blotting
IEVH
20
0.45 μm
Videos
SDS
Documents

Documents