Learn More
Cytiva Immobiline™ DryStrip Kit and Components
Catalog No. 45000417
Change view
Click to view available options
Type:
Sample Cup
Catalog No. Type
45000417 Sample Cup
1 options
Catalog No. 45000417 Supplier Cytiva Supplier No. 18100435/DUP
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Precast Immobiline DryStrip gels for running the first dimension of a 2-D electrophoresis experiment with the Multiphor II electrophoresis system
Run up to 12 Immobiline DryStrip strips simultaneously with up to 100μL on each Immobiline DryStrip.
  • The instruction manual presents all necessary information for the entire method for horizontal 2-D electrophoresis. By appropriate adaptations, the SDS electrophoresis can also be performed using vertical electrophoresis equipment
  • Immobiline DryStrip kit facilitates sample application and running and equilibration of Immobiline DryStrip for the first dimension in 2-D (two-dimensional) electrophoresis
  • Isoelectric focusing with an immobilized pH gradient (IPG), makes true isoelectric focusing possible and significantly improves the reproducibility of the spot distribution along the pH gradient axis of 2-D maps
  • Individual components are also available separately
Specifications
For Use With (Equipment) Multiphor II Electrophoresis System
Type Sample Cup