missing translation for 'onlineSavingsMsg'
Learn More

Iminodiacetic Acid 98.0+%, TCI America™

Catalog No. I0006500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0006500G 500 g
I000625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0006500G Supplier TCI America Supplier No. I0006500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 142-73-4
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.103
MDL Number MFCD00004280
InChI Key NBZBKCUXIYYUSX-UHFFFAOYSA-N
Synonym iminodiacetic acid, 2,2'-iminodiacetic acid, glycine, n-carboxymethyl, aminodiacetic acid, diglycocoll, diglykokoll, diglycin, iminodiethanoic acid, n-carboxymethyl glycine, 2,2'-azanediyldiacetic acid
PubChem CID 8897
ChEBI CHEBI:24786
IUPAC Name 2-(carboxymethylamino)acetic acid
SMILES C(C(=O)O)NCC(=O)O

Specifications

Melting Point 250°C
Color White
Quantity 500 g
Formula Weight 133.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Iminodiacetic Acid
EINECSNumber (2)-1211
RTECSNumber AI2975000
TSCA Yes