missing translation for 'onlineSavingsMsg'
Learn More

Imidurea, 26-28% nitrogen

Catalog No. AC268702500
Change view
Click to view available options
Quantity:
100 g
250 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC268702500 250 g Glass bottle
AC268701000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC268702500 Supplier Thermo Scientific Chemicals Supplier No. 268702500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 39236-46-9 | C11H16N8O8 | 388.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39236-46-9
Molecular Formula C11H16N8O8
Molecular Weight (g/mol) 388.29
MDL Number MFCD00221482
InChI Key ZCTXEAQXZGPWFG-UHFFFAOYSA-N
Synonym imidazolidinyl urea, imidurea, germall 115, 1,1'-methylenebis 3-3-hydroxymethyl-2,5-dioxoimidazolidin-4-yl urea, imidazolinidyl urea, imidurea nf, methanebis n,n'-5-ureido-2,4-diketotetrahydroimidazole-n,n-dimethylol, n,n-methylenebis n'-1-hydroxymethyl-2,5-dioxo-4-imidazolidinyl urea, 1,1'-methylenebis 3-3-hydroxymethyl-2,5-dioxo-4-imidazolidinyl urea
PubChem CID 38258
ChEBI CHEBI:51805
IUPAC Name 1-[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-3-[[[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]carbamoylamino]methyl]urea
SMILES C(NC(=O)NC1C(=O)NC(=O)N1CO)NC(=O)NC2C(=O)NC(=O)N2CO

Specifications

pH 6.0 to 7.5 (1% aq. soln.)
Loss on Drying 3.0 % max. (48 hrs) (vacuum, P2O5)
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 250 g
Merck Index 15, 4957
Solubility Information Solubility in water: soluble
Formula Weight 388.29
Percent Purity 26-28% nitrogen
Physical Form Liquid
Chemical Name or Material Imidurea, 26-28% nitrogen
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
EINECSNumber 254-372-6
RTECSNumber YT7365000
TSCA TSCA

RUO – Research Use Only