missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Imidazole, 0.5M buffer soln., pH 7.5

Catalog No. AAJ63496AK
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63496AK 250 mL
AAJ63496AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63496AK Supplier Thermo Scientific Chemicals Supplier No. J63496AK

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 288-32-4 | C3H4N2 | 68.08 g/mol

Imidazole is used as a chelator for the binding of various divalent cations, and is also used to elute tagged proteins bound to Ni ions attached to the surface of beads in the chromatography column.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Imidazole is used as a chelator for the binding of various divalent cations, and is also used to elute tagged proteins bound to Ni ions attached to the surface of beads in the chromatography column.

Solubility
Fully miscible.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 288-32-4
Molecular Formula C3H4N2
Molecular Weight (g/mol) 68.08
MDL Number MFCD00005183
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
IUPAC Name 1H-imidazole
SMILES N1C=CN=C1

Specifications

Concentration 0.5 M
pH 7.5
Physical Form Liquid
Quantity 250 mL
Chemical Name or Material Imidazole
Recommended Storage Keep cold

RUO – Research Use Only