missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Imidazo[1,2-b]pyridazine, 98%, Thermo Scientific™
Spécifications
Spécifications
| Nom ou substance chimique | Imidazo[1,2-b]pyridazine |
| Spectre infrarouge | Authentic |
| Fourchette de pourcentages d’essai | 97.5% min. (GC) |
| Emballage | Glass Bottle |
| Quantité | 1g |
| Renseignements sur la solubilité | Solubility in water: .. Other solubilities: soluble in dmf |
| Poids de formule | 119.12 |
| Pourcentage de pureté | 98% |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present a |
RUO – Research Use Only