missing translation for 'onlineSavingsMsg'
Learn More

Acide imidazo[1,2-a]pyridine-3-carboxylique, 97%, Thermo Scientific™

Numéro de catalogue. CC25401CB
Modifier l'affichage
Click to view available options
Quantité:
250 mg
Emballage:
Bouteille en verre ambrée
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC25401CB 250 mg Bouteille en verre ambrée
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC25401CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC25401CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 6200-60-8
Formule moléculaire C8H6N2O2
Poids moléculaire (g/mol) 162.15
Numéro MDL MFCD01720458
Clé InChI DOGXPDFZEQXZDS-UHFFFAOYSA-N
Synonyme imidazo 1,2-a pyridine-3-carboxylic acid, imidazo 1,2-a pyridine-3-carboxylicacid, pubchem19678, wln: t56 an dnj bvq, 3-carboxyimidazo 1,2-a pyridine, h-imidazo 1,2-a pyridine-3-carboxylic acid, 4-hydroimidazo 1,2-a pyridine-3-carboxylic acid, 1h-imidazo 1,2-a pyridine-3-carboxylic acid
PubChem CID 73142
Nom de l’IUPAC Acide imidazo[1,2-A]pyridine-3-carboxylique
SOURIRES OC(=O)C1=CN=C2C=CC=CN12

Spécifications

Nom ou substance chimique Imidazo[1, 2-a]pyridine-3-carboxylic acid
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Bouteille en verre ambrée
Quantité 250 mg
Poids de formule 162.15
Pourcentage de pureté 97%
Hazard Category Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.