missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva PuReTaq Ready-To-Go™ PCR Beads
Description
- puReTaq DNA polymerase and high-purity reagents provide robust performance with very low levels of contaminating DNA
- Manufacturing process locks in performance for excellent reproducibility with less risk of pipetting errors and contamination
- Long-term ambient-temperature stability simplifies shipping and removes any need for freezer storage
- Verified for real-time PCR
- Ready-To-Go means simply add template DNA and primers and cycle
Specifications
Specifications
| Content And Storage | Room temperature stable beads containing ∼ 2.5 units of recombinant PuReTaq DNA Polymerase, dATP, dCTP, dGTP, dTTP, stabilizers, BSA and reaction Buffer. |
| Format | 0.2mL Tube with hinged cap |
| Quantity | 1 Kit |
| For Use With (Application) | Standard PCR |
| No. of Reactions | 96 |
| Volume | 200μL |