missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva GenomiPhi V2 DNA Amplification Kit
Amplify whole genomes with high efficiency and representation. Cytiva Life Sciences™ illustra™ GenomiPhi V2 DNA Amplification Kit provides whole genome amplification with yields of 4 to 7μg.
$336.00 - $4,293.00
Specifications
| Content And Storage | Store the kit at -70°C.The enzyme mix must be stored at -70°C; all other components may be stored at -20°C. Thaw components on ice and maintain at 0°C to 4°C during handling. |
|---|---|
| Description | Illustra GenomiPhi V2 DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 4 to 7 μg yield from nanogram amounts of DNA sample. |
| For Use With (Application) | Genomic DNA Amplification |
| Product Type | illustra GenomiPhi V2 DNA Amplification Kit |
| Size | > 10 kb |
Includes: Sample buffer, Reaction buffer, Enzyme mix, and Lambda control DNA (10ng/μL)
| Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity & Availability | |||||
|
45001221
|
Cytiva
25660030 |
25 reactions |
Each for $336.00
|
|
|||||
|
45001222
|
Cytiva
25660031 |
100 reactions |
Each for $929.00
|
|
|||||
|
45001223
|
Cytiva
25660032 |
500 reactions |
Each for $4,293.00
|
|
|||||
Description
- Starting material can be purified DNA or nonpurified cell lysates
- Simple protocol provides typical yields of 4 to 7μg DNA and average product lengths >10kb
- Less hands-on time compared to traditional isolation methods
- Representative amplification of the whole genome
- Automation-friendly protocol with great reproducibility
- High-quality DNA for array CGH, high-throughput genotyping, DNA archival, PCR, restriction enzyme digestion, hybridization, and cloning
- Contains all of the components necessary for genomic DNA preparation by isothermal strand displacement amplification
Specifications
| Store the kit at -70°C.The enzyme mix must be stored at -70°C; all other components may be stored at -20°C. Thaw components on ice and maintain at 0°C to 4°C during handling. | |
| Genomic DNA Amplification | |
| > 10 kb |
| Illustra GenomiPhi V2 DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 4 to 7 μg yield from nanogram amounts of DNA sample. | |
| illustra GenomiPhi V2 DNA Amplification Kit | |
| 1.5-2 h |