missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva GenomiPhi V2 DNA Amplification Kit
Amplify whole genomes with high efficiency and representation. Cytiva Life Sciences™ illustra™ GenomiPhi V2 DNA Amplification Kit provides whole genome amplification with yields of 4 to 7μg.
Supplier: Cytiva 25660030
Includes: Sample buffer, Reaction buffer, Enzyme mix, and Lambda control DNA (10ng/μL)
Description
- Starting material can be purified DNA or nonpurified cell lysates
- Simple protocol provides typical yields of 4 to 7μg DNA and average product lengths >10kb
- Less hands-on time compared to traditional isolation methods
- Representative amplification of the whole genome
- Automation-friendly protocol with great reproducibility
- High-quality DNA for array CGH, high-throughput genotyping, DNA archival, PCR, restriction enzyme digestion, hybridization, and cloning
- Contains all of the components necessary for genomic DNA preparation by isothermal strand displacement amplification
Chemical Identifiers
| Store the kit at -70°C.The enzyme mix must be stored at -70°C; all other components may be stored at -20°C. Thaw components on ice and maintain at 0°C to 4°C during handling. | |
| Genomic DNA Amplification | |
| > 10 kb |
| Illustra GenomiPhi V2 DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 4 to 7 μg yield from nanogram amounts of DNA sample. | |
| illustra GenomiPhi V2 DNA Amplification Kit | |
| 1.5-2 h |
Specifications
| Store the kit at -70°C.The enzyme mix must be stored at -70°C; all other components may be stored at -20°C. Thaw components on ice and maintain at 0°C to 4°C during handling. | |
| Genomic DNA Amplification | |
| 25 reactions | |
| > 10 kb |
| Illustra GenomiPhi V2 DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 4 to 7 μg yield from nanogram amounts of DNA sample. | |
| 225μL Sample Buffer, 225μL Reaction Buffer, 25μL Enzyme mix, 20μL Control DNA (lambda) | |
| illustra GenomiPhi V2 DNA Amplification Kit | |
| 1.5-2 h |