missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL1F10 (IL-38) Mouse anti-Human, Clone: H127C, MilliporeSigma™
Mouse Monoclonal Antibody
$284.60 - $731.13
Specifications
| Antigen | IL1F10 (IL-38) |
|---|---|
| Clone | H127C |
| Concentration | 1.0 mg/mL |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
MABF275425
|
N/A | 25 μg |
Each for $284.60
|
|
|||||
|
MABF2754100
|
N/A | 100 μg |
Each for $731.13
|
|
|||||
Description
IL1F10 (IL-38) Monoclonal antibody specifically detects IL1F10 (IL-38) in Human samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blotting.Specifications
| IL1F10 (IL-38) | |
| 1.0 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
| IgG2b κ | |
| Protein G purified |
| H127C | |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| His-tagged full-length recombinant human Interleukin-1 family member 10 (IL1F10). | |
| Primary | |
| Stable for 1 year at +2°C to +8°C from date of receipt. |