missing translation for 'onlineSavingsMsg'
Learn More

IL1F10 (IL-38) Mouse anti-Human, Clone: H127C, MilliporeSigma™

Catalog No. MABF275425
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF275425 25 μg
MABF2754100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF275425 Supplier MilliporeSigma Supplier No. MABF275425UG

Mouse Monoclonal Antibody

IL1F10 (IL-38) Monoclonal antibody specifically detects IL1F10 (IL-38) in Human samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blotting.

Specifications

Antigen IL1F10 (IL-38)
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone H127C
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen His-tagged full-length recombinant human Interleukin-1 family member 10 (IL1F10).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for 1 year at +2°C to +8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less