missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL1F10 (IL-38) Mouse anti-Human, Clone: H127C, MilliporeSigma™
Description
IL1F10 (IL-38) Monoclonal antibody specifically detects IL1F10 (IL-38) in Human samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications
Specifications
| Antigen | IL1F10 (IL-38) |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | H127C |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. |
| Host Species | Mouse |
| Immunogen | His-tagged full-length recombinant human Interleukin-1 family member 10 (IL1F10). |
| Purification Method | Protein G purified |
| Show More |