missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ IL-5 ELISPOT Antibody Pair, Human

Catalog No. ELI006H
Change view
Click to view available options
Quantity:
96-well Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ELI006H 96-well Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ELI006H Supplier MilliporeSigma™ Supplier No. ELI006H

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Protein Interaction Antibody Pairs

Includes:

  • Human IL-5 Capture Antibody, clone TRFK5. Quantity: 500 μg.
  • Human IL-5 Detection Antibody, clone JES1-5A10. Quantity: 100 μg.
  • Streptavidin Alkaline Phosphatase. Quantity: 55 μL.

Specifications

Classification Monoclonal
Concentration 1.0mg/mL (Capture antibody), 0.5mg/mL (Biotinylated detection antibody)
Host Species Rat
Quantity 96-well Plates
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for up to six months
Product Type TRFK5 (Capture), JES1-5A10 (Detection)
Form Purified