missing translation for 'onlineSavingsMsg'
Learn More

PeproTech IL-16 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Numéro de catalogue. 500P06BT25
Encompass
Change view
Click to view available options
Quantity:
25 μg
50 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
500P06BT25 25 μg
500P06BT50 50 μg
502718719 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. 500P06BT25 Fournisseur PeproTech Code fournisseur 500P06BT25UG
Il en reste null

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: PDLNSSTDSA ASASAASDVS VESTAEATVC TVTLEKMSAG LGFSLEGGKG SLHGDKPLTI NRIFKGAASE QSETVQPGDE ILQLGGTAMQ GLTRFEAWNI IKALPDGPVT IVIRRKSLQS KETTAAGDS. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human IL-16. Anti-Human IL-16-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human IL-16 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human IL-16 (500-P06) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human IL-16. Western Blot: To detect hIL-16 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hIL-16 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P06BT-1MG will be provided as 2 x 500 μg

The protein encoded by IL-16 is a pleiotropic cytokine that functions as a chemoattractant, a modulator of T cell activation, and an inhibitor of HIV replication. The signaling process of this cytokine is mediated by CD4. The product of IL-16 undergoes proteolytic processing, which is found to yield two functional proteins. The cytokine function is exclusively attributed to the secreted C-terminal peptide, while the N-terminal product may play a role in cell cycle control. Caspase 3 is reported to be involved in the proteolytic processing of this protein. Alternate splicing results in multiple transcript variants.

Spécifications

Antigen IL-16
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Biotin
Formulation PBS with no preservative
Gene IL16
Gene Accession No. Q14005
Gene Alias FLJ16806; FLJ42735; FLJ44234; H-IL-16; il 16; Il16; IL-16; ILN; Interleukin; interleukin 16; Interleukin16; Interleukin-16; LCF; Lymphocyte chemoattractant factor; M-IL-16; mKIAA4048; neuronal interleukin 16; NIL16; PRIL16; prIL-16; prointerleukin 16; Pro-interleukin-16
Gene Symbols IL16
Host Species Rabbit
Immunogen E.coli-derived Recombinant Human IL-16 (129 a.a.).
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3603
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Afficher plus de résultats Afficher moins de résultats