missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-11 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Numéro de catalogue. 500P01BT50
Encompass
Modifier l'affichage
Click to view available options
Quantité:
25 μg
50 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
500P01BT50 50 μg
500P01BT25 25 μg
502718727 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. 500P01BT50 Fournisseur Invitrogen™ Code fournisseur 500P01BT50UG
Saison des économies
Afficher toutes les promotions

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: MPGPPPGPPR VSPDPRAELD STVLLTRSLL ADTRQLAAQL RDKFPADGDH NLDSLPTLAM SAGALGALQL PGVLTRLRAD LLSYLRHVQW LRRAGGSSLK TLEPELGTLQ ARLDRLLRRL QLLMSRLALP QPPPDPPAPP LAPPSSAWGG IRAAHAILGG LHLTLDWAVR GLLLLKTRL. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human IL-11. Anti-Human IL-11-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human IL-11 by sandwich ELISA (using 100 μL/well) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human IL-11 (500-P01) as a capture antibody, allows the detection of at least 2000-4000 pg/mL of Recombinant Human IL-11 Western Blot: To detect Human IL-11 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible development reagents the detection limit for Recombinant Human IL-11 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P01BT-1MG will be provided as 2 x 500 μg

Interleukins (ILs) are a large group of cytokines that are produced mainly by leukocytes, although some are produced by certain phagocytes and auxiliary cells. ILs have a variety of functions, but most function to direct other immune cells to divide and differentiate. Each IL acts on a specific, limited group of cells through a receptor specific for that IL.

Spécifications

Antigène IL-11
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugué Biotin
Formule PBS with no preservative
Gène IL11
Numéro d’acquisition du gène P20809
Alias du gène adipogenesis inhibitory factor; AGIF; il 11; IL10RA; Il11; IL-11; IL11 inhibitor; IL-11ra; ILN; Interleukin; interleukin 11; Interleukin11; interleukin-11; MGC2146; Oprelvekin
Symboles de gène IL11
Espèces hôtes Rabbit
Immunogène E.coli-derived Recombinant Human IL-11.
Méthode de purification Antigen affinity chromatography
Quantité 50 μg
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 3589
Espèces cibles Human
Contenu et stockage -20°C
Type de produit Antibody
Forme Lyophilized
Afficher plus de résultats Afficher moins de résultats