Learn More
Invitrogen™ IL-1 beta Polyclonal Antibody, Biotin, eBioscience™, Invitrogen™

Description
Description: The biotinylated polyclonal rabbit antibody reacts with mouse interleukin-1beta (IL-1beta), a 17 kDa factor secreted primarily by monocytes. IL-1 has effects on T-helper cells, inducing IL-2 secretion and expression of IL-2 receptors. IL-1 has effects on B cells, promoting cell proliferation and immunoglobulin synthesis. Applications Reported: Biotin anti-mouse interleukin-1 beta (IL-1 beta) has been reported for use as the detection antibody for a mouse IL-1 beta ELISA. Applications Tested: The biotinylated polyclonal rabbit anti-mouse IL-1 beta antibody has been tested as the detection antibody in a sandwich ELISA for analysis of mouse IL-1 beta in combination with the unlabelled B122 (14-7012) antibody for capture and recombinant mouse IL-1 beta (14-8012) as the standard. A suitable range of concentrations of this antibody for ELISA detection is 1.0 - 6.0 μg/mL. A standard curve consisting of doubling dilutions of the recombinant standard over the range of 2000 pg/mL - 15 pg/mL should be included in each ELISA plate. Filtration: 0.2 μm post-manufacturing filtered.
Spécifications
Spécifications
| Antigène | IL-1 beta |
| Applications | ELISA |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugué | Biotin |
| Formule | PBS with 0.09% sodium azide; pH 7.2 |
| Gène | IL1B |
| Numéro d’acquisition du gène | P10749 |
| Alias du gène | catabolin; cytokine; Hematopoietin 1 (H1); IFN beta inducing factor; il 1b; IL 1β; IL-1; IL1 B; IL-1 beta; IL-1 precursor; IL1B; IL-1B; IL1B1; IL1beta; IL-1beta; IL1-BETA; IL1F2; IL1β; ILN; Interleukin; interleukin 1 beta; Interleukin 1 beta precursor; interleukin 1, beta; interleukin 1, beta 1; interleukin 1-beta; interleukin*1*beta; Interleukin1 beta; interleukin-1 beta; interleukin-1 beta precursor; interleukin-1 beta precursor (AA -113 to 153); interleukin-1 beta proprotein; interleukin-1b; interleukin-1beta; interleukin-beta; LAF; lymphocyte proliferation-potentiating factor; Osteoclast activating factor (OAF); preinterleukin 1 beta; Pro interleukin 1 beta; prointerleukin-1 beta; pro-interleukin-1-beta |
| Symboles de gène | IL1B |
| Afficher plus de résultats |
For Research Use Only.