missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IKA™ Shaker numérique MS 3
Description
The IKA MS 3 Digital Shaker is a compact, universal shaker suitable for room temperature shaking tasks in the QuantiGene Plex assay protocol.
The IKA MS 3 digital shaker has the following features:
- Versatile configuration—a wide range of attachments and attachment detection
- Precision—includes a timer with countdown function
- Ease of operation—has a continuous or touch operation (with standard attachment)
- Flexible operating modes:
- Mode A (safe mode with attachment detection)—the maximum speed of 3,000 rpm is only reached with the standard attachment in touch mode. When using other attachments, the speed is limited to 1,300 rpm.
- Mode B (without attachment detection)—a speed of 3,000 rpm is possible with all attachments
- Stable action in all speed ranges
- Sturdy construction—zinc die cast casing
Spécifications
Spécifications
| Plage de vitesse | 0 à 3000 tr/min |
| Type de produit | Shaker numérique |
| Certifications/Conformité | Certificat d’analyse (COA) |
| Modèle | IKA Shaker numérique MS 3 |
| Affichage | Numérique |
| Gamme de produit | IKA |
| Quantité | 1 shaker |
Usage exclusivement réservé à la recherche. Ne pas utiliser dans des procédures de diagnostic.