Learn More
Invitrogen™ Mouse IgM Monoclonal Antibody (eB121-15F9), PE-Cyanine7, eBioscience™

Description
Description: The eB121-15F9 monoclonal antibody reacts with the mu heavy chain of mouse IgM. It does not react with other classes of mouse immunoglobulin including IgD, IgG or IgA. IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells. Fluorochrome conjugated eB121-15F9 can be used as a detection secondary for mouse IgM. Applications Reported: This eB121-15F9 antibody has been reported for use in flow cytometric analysis. Applications Tested: This eB121-15F9 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-822-49) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333-54) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore...
Spécifications
Spécifications
| Antigène | Mouse IgM |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | eB121-15F9 |
| Concentration | 0.2 mg/mL |
| Conjugué | PE-Cyanine7 |
| Formule | PBS with 0.09% sodium azide; pH 7.2 |
| Gène | Ighm |
| Numéro d’acquisition du gène | P01872 |
| Alias du gène | AGM1; AI326478; Igh6; Igh-6; Ighm; Igh-M; Igm; immunoglobulin heavy constant mu; MU; muH; VH |
| Afficher plus de résultats |