missing translation for 'onlineSavingsMsg'
Learn More

Igepal Ca - 630 Non-Ionic Detergent MP Biomedicals

Catalog No. ICN19859650
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19859650 50 mL
ICN19859680 100 mL
ICN19859690 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19859650 Supplier MP Biomedicals Inc Supplier No. 0219859650

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9036-19-5
Molecular Formula (C2H4O)x·C14H22O
Synonym Polyethylene glycol octylphenol ether

Specifications

Density 1.06 g/mL
Quantity 50 mL
Chemical Name or Material Igepal Ca - 630, Non-Ionic Detergent