missing translation for 'onlineSavingsMsg'
Learn More

IGEPAL™ CA-630 , MilliporeSigma™ Supelco™

Catalog No. 111020016
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020016 50 mL
111018967 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020016 Supplier Merck Emd Millipore Supplier No. 5674150MLF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

IGEPAL™ CA-630 is a nonionic, non-denaturing detergent suitable for solubilization, isolation and purification of membrane protein complexes. It is chemically indistinguishable from Nonidet P-40.

Specifications

CAS 9002-93-1
Density 1.06 g/mL (at 20°C)
Quantity 50 mL
Linear Formula (C2H4O)nC14H22O
Refractive Index n20/D 1.492
Synonym Nonidet(TM) P 40; Nonylphenyl-polyethylenglycol; Octylphenoxy poly(ethyleneoxy)ethanol, branched; Octylphenoxy poly(ethyleneoxy)ethanol, branched
Molecular Weight (g/mol) 603.0

IGEPAL is a registered trademark of Rhodia Operations