Learn More
Invitrogen™ Mouse IgD Monoclonal Antibody (11-26c (11-26)), APC-eFluor™ 780, eBioscience™

Description
Description: The 11-26c monoclonal antibody reacts with the delta heavy chain of mouse IgD. It does not react with other classes of mouse immunoglobulin including IgA, IgG or IgM. IgD is expressed by peripheral mature B cells. 11-26c does not activate B cells. Applications Reported: This 11-26c (11-26) antibody has been reported for use in flow cytometric analysis. Applications Tested: This 11-26c (11-26) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. APC-eFluor® 780 emits at 780 nm and is excited with the Red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance...
Specifications
Specifications
| Antigène | Mouse IgD |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 11-26c (11-26) |
| Concentration | 0.2 mg/mL |
| Conjugué | APC-eFluor 780 |
| Formule | PBS with 0.09% sodium azide; pH 7.2 |
| Gène | Ighd |
| Numéro d’acquisition du gène | P01881 |
| Alias du gène | IgD; Igh-5; IGHD; immunoglobulin heavy constant delta |
| Show More |