Recombinant human NRAS (Q61K) (1-186) expressed in E. coli cells using an N-terminal Avi-His tag and enzymatically biotinylated in vitro prior to the final chromatography purification. NRAS (Q61K) is constitutively active.
Numéro de catalogue.
502573328
Fournisseur
SignalChem BiotechCode fournisseur
R0732HB1000
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Recombinant human NRAS (Q61K) (1-186) expressed in E. coli cells using an N-terminal Avi-His tag and enzymatically biotinylated in vitro prior to the final chromatography purification. NRAS (Q61K) is constitutively active.