missing translation for 'onlineSavingsMsg'
Learn More

Idebenone 98.0+%, TCI America™

$190.13 - $971.17

Chemical Identifiers

CAS 58186-27-9
Molecular Formula C19H30O5
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I08481G
View Documents
TCI America
I08481G
1g
Each for $190.13
Only null left
 
I08485G
View Documents
TCI America
I08485G
5g
Each for $316.06
Only null left
 
I084825G
View Documents
TCI America
I084825G
25g
Each for $971.17
Only null left
 
Description

Description

Specifications

Chemical Identifiers

58186-27-9
C19H30O5

Specifications

58186-27-9
53°C
338.44
Undesignated
Idebenone
1g
C19H30O5
Crystalline Powder at 20°C
≥98.0% (HPLC,T)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'