missing translation for 'onlineSavingsMsg'
Learn More

Idebenone 98.0+%, TCI America™

Catalog No. I08481G
Change view
Click to view available options
Quantity:
1g
5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08481G 1g
I08485G 5g
I084825G 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I08481G Supplier TCI America Supplier No. I08481G

Chemical Identifiers

CAS 58186-27-9
Molecular Formula C19H30O5

Specifications

Quantity 1g
Melting Point 53°C
Formula Weight 338.44
Physical Form Crystalline Powder at 20°C
Color Undesignated
Purity ≥98.0% (HPLC,T)
Chemical Name or Material Idebenone