missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Idebenone, 98.5%, Spectrum™ Chemical
$304.62 - $24902.26
Chemical Identifiers
| CAS | 58186-27-9 |
|---|---|
| Molecular Formula | C19H30O5 |
| Molecular Weight (g/mol) | 338.44 |
| InChI Key | JGPMMRGNQUBGND-UHFFFAOYSA-N |
| IUPAC Name | 2-(10-hydroxydecyl)-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione |
| SMILES | COC1=C(OC)C(=O)C(CCCCCCCCCCO)=C(C)C1=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18611010
|
Spectrum Chemical Mfg Cor
I20165GM |
5 g | Amber Glass Bottle |
Each for $304.62
|
|
||||
|
18611009
|
Spectrum Chemical Mfg Cor
I201625GM |
25 g | Amber Glass Bottle |
Each for $1,000.68
|
|
||||
|
18611008
|
Spectrum Chemical Mfg Cor
I2016100GM |
100 g | Amber Glass Bottle |
Each for $3,520.29
|
|
||||
|
18611007
|
Spectrum Chemical Mfg Cor
I20161KGBL |
1 kg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 58186-27-9 | |
| 338.44 | |
| 2-(10-hydroxydecyl)-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione |
| C19H30O5 | |
| JGPMMRGNQUBGND-UHFFFAOYSA-N | |
| COC1=C(OC)C(=O)C(CCCCCCCCCCO)=C(C)C1=O |
Specifications
| 58186-27-9 | |
| 52.0° to 55.0°C | |
| Amber Glass Bottle | |
| 5 g | |
| COC1=C(OC)C(=O)C(CCCCCCCCCCO)=C(C)C1=O | |
| 338.44 | |
| Reagent |
| 1 | |
| 0.02 | |
| C19H30O5 | |
| JGPMMRGNQUBGND-UHFFFAOYSA-N | |
| 2-(10-hydroxydecyl)-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione | |
| 0.985 |
CAUTION: For use by qualified personnel for research and development or other authorized use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.