missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Idebenone, 98.5%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor I2016100GM
Specifications
| 58186-27-9 | |
| Amber Glass Bottle | |
| 100 g | |
| COC1=C(OC)C(=O)C(CCCCCCCCCCO)=C(C)C1=O | |
| 338.44 |
| 52.0° to 55.0°C | |
| C19H30O5 | |
| JGPMMRGNQUBGND-UHFFFAOYSA-N | |
| 2-(10-hydroxydecyl)-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione | |
| Reagent |
Chemical Identifiers
| 58186-27-9 | |
| 338.44 | |
| 2-(10-hydroxydecyl)-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione |
| C19H30O5 | |
| JGPMMRGNQUBGND-UHFFFAOYSA-N | |
| COC1=C(OC)C(=O)C(CCCCCCCCCCO)=C(C)C1=O |
CAUTION: For use by qualified personnel for research and development or other authorized use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.