missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Idebenone

Numéro de catalogue. p-7214796
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AC466770010 1 g
AC466770050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC466770010 Fournisseur Thermo Scientific Chemicals Code fournisseur 466770010
Il en reste null

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 58186-27-9
Molecular Formula C19H30O5

Spécifications

Quantity 1 g
Solubility Solubility aq. soln.: insoluble. Other solubilities: slightly soluble in methanol
Melting Point 52.0°C to 55.0°C
Formula Weight 338.44
InChI Key JGPMMRGNQUBGND-UHFFFAOYSA-N
SMILES COC1=C(OC)C(=O)C(CCCCCCCCCCO)=C(C)C1=O
IUPAC Name 2-(10-hydroxydecyl)-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione
Molecular Weight (g/mol) 338.44
Chemical Name or Material Idebenone

RUO – Research Use Only