missing translation for 'onlineSavingsMsg'
Learn More

Abbott™ ID NOW RSV Control Swab

Catalog No. 23043633
Encompass
Change view
Click to view available options
Quantity:
20/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23043633 20/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Refer to Package Insert for list of warnings
Catalog No. 23043633 Supplier Abbott™ Supplier No. 435080

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Refer to Package Insert for list of warnings

Includes

10 posive controls swabs, 10 negative controls swabs, Package Insert

Highly sensitive rapid diagnostic tests for RSV are needed to help guide management in the clinical setting. Combines the benefits of speed and accuracy on an easy to use system.

RSV control swab to be used with Abbott ID NOW RSV test kit.

Certifications

CLIA-Moderate

Specifications

Certifications/Compliance CLIA Moderate
Content And Storage Room Temperature
CPT Code 87801
CE Marker Yes
Clia Complexity Moderate
Detectable Analytes Respiratory Syncytial Virus
Sensitivity Direct NPS: 98.6%, NPS eluted in VTM: 98.6%
Type Quality Controls - RSV
For Use With (Application) Quality control for diagnosis of RSV
Material Cardboard
Packaging Type Cardboard Box
Quantity 20/Pk.
State at Room Temperature Stable: Dry swab
Test Time 13 min. or less
Show More Show Less