missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Ibrutinib

Numéro de catalogue. 681350
Modifier l'affichage
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
681350 50 mg
681310 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 681350 Fournisseur Tocris Bioscience™ Code fournisseur 6813/50
Il en reste null

Potent and selective BTK inhibitor (IC50= 0.5 nM)

Ibrutinib is a potent and selective BTK inhibitor (IC50= 0.5 nM). Selective for BTK against a screening panel of kinase enzymes. Inhibits autophosphorylation of BTK, phosphorylation of PLCγ, and phosphorylation of ERK (IC50 of 11 nM, 29 nM and 13 nM, respectively). Also inhibits ErbB4 (IC50 = 0.25-3.4 nM). Induces cytotoxicity of chronic lymphocytic leukemia (CLL) cells. Reduces clinical arthritis scores in a mouse model.

Spécifications

Chemical Name or Material 1-[(3R)-3-[4-Amino-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl]-1-piperidinyl]-2-propen-1-one
CAS 936563-96-1
Synonym Agammaglobulinaemia tyrosine kinase, AGMX1, ATKIMD1, B-cell progenitor kinase, BPK, Bruton agammaglobulinemia tyrosine kinase, Bruton tyrosine kinase, dominant-negative kinase-deficient Brutons tyrosine kinase, tyrosine-protein kinase
Quantity 50 mg
Molecular Formula C25H24N6O2
Formula Weight 440.5
Purity 98%