Learn More
Invitrogen™ iBright™ System Tray Adapter for E-Gel™ Agarose Gels
Description
Invitrogen iBright Tray Adapters for E-Gel Agarose Gels are imaging accessories designed to optimize the image capture of Invitrogen E-Gel Agarose Gels.
Features of Invitrogen iBright Tray Adapters for E-Gel Agarose Gels include:
• The bottom component centers the E-Gel agarose gels on the iBright transilluminator box facilitating optimized zoom
• The top component covers labels and bar codes on the E-Gel cassette to reduce stray light during image capture and improve sample signal optimization
• Compatible with all iBright models (CL750, CL1000, CL1500, FL1000, and FL1500)
• Compatible with 11- to 96-well E-Gel agarose (Cat. No. A56600 for 11-/22-well, Cat. No. A56599 for 48-/96-well)
Spécifications
Spécifications
| Product Line | iBright |
| For Use With (Application) | Chemiluminescent Western Blot Imaging, DNA Gel Imaging, Protein Gel Imaging |
| Shipping Condition | Room Temperature |
| Quantity | 1 Adaptor |
| Product Type | E-Gel Tray Adapter, 48-/96-well |