Learn More
Invitrogen™ iBind Flex PLUS Solution Kit
Description
The iBind Flex PLUS Solution Kit increases signal sensitivity, requires less antibody per run, and simplifies western blotting immunoprocessing when used with the iBind Flex Western System. The iBind Flex PLUS Solution Kit comes with two ready to use solutions, iBind Flex PLUS Solution and iBind Flex PLUS Antibody Diluent.
- The iBind Flex PLUS Solution comes ready to use with no preparation required. It is used to prewet the iBind Flex card prior to the run and acts as the block and wash buffer for the membrane processing, helping wash away unbound primary and secondary antibodies.
- The iBind Flex PLUS Antibody Diluent is used to dilute the primary and secondary antibodies. In most experiments, the primary antibody can be diluted to the same recommended concentration as used for traditional blotting, however only 2 mL (mini blot), 4 mL (midi blot) or 0.7 mL (strips) of prepared primary antibody is required with iBind Flex, saving up to 80% of primary antibody consumption. The iBind Flex PLUS Antibody Diluent is specially formulated with bulking agents and stabilizing molecules to improve antibody binding with the target antigen to increase signal intensity.
During a run on the iBind Flex, each solution is released from the iBind Flex device wells through sequential lateral flow (SLF) and are wicked over the iBind Flex card towards the stack region, incubating the membrane with each reagent. The glass fiber matrix of the iBind Flex card allows for homogenous and consistent flow of the solutions to the membrane, increasing the antigen-antibody interaction.
The iBind Flex PLUS Solution Kit, together with the iBind Flex Western Device (Cat. No. SLF2000) and iBind Flex Cards (Cat. No. SLF2010), bring automation and simplicity to western blotting with efficient and hands-free immunoprocessing in as little as 3 hours. Compared to manual methods, the iBind Flex western system requires less primary antibody, enables reproducible results, and is compatible with downstream detection protocols.
Each iBind Flex PLUS Solution Kit contains sufficient reagents for 10 immunoprocessing reactions with flexibility of each reaction supporting 1 midi blot, 2 mini blots, or 6 vertically cut strips.
For iBind Devices, use the iBind PLUS Solution Kit. For fluorescent applications, use the iBind Fluorescent Detection (FD) Solution Kit (Cat. No. SLF1019) or iBind Flex Fluorescent Detection (FD) Solution Kit (Cat. No. SLF2019).
Spécifications
Spécifications
| Compatibilité des membranes | Nitrocellulose, PVDF |
| À utiliser avec (équipement) | iBind Flex Western Device |
| Contenu et stockage | iBind Flex PLUS Solution, iBind Flex PLUS Antibody Diluent Store at 2°C to 8°C |
| Méthode de détection | Colorimetric, Chromogenic, Chemiluminescence |
| Type de produit | Flex Solution Kit |
| Nb de réactions | 40 Midi Blots or 80 Mini Blots |
| Compatibilité avec le gel | Mini Precast Protein Gels, Midi Precast Protein Gels |
| Gamme de produit | iBind |
| Quantité | 4 Kits |
For Research Use Only. Not for use in diagnostic procedures.