missing translation for 'onlineSavingsMsg'
Learn More

IBI Scientific™ Blot Washer Trays

Catalog No. 361004445
Change view
Click to view available options
Type:
Large Blot Tray
Mini Blot Tray
Standard Blot Tray
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
361004445 Large Blot Tray
361004443 Mini Blot Tray
361004444 Standard Blot Tray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 361004445 Supplier IBI Scientific™ Supplier No. ACCBW0004

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with Blot Washer system, Belly Dancer Shaker and Hi/Lo Rocker

  • Comes with a lid
  • Available in MINI, Standard and Large

Specifications

Color Clear
For Use With (Equipment) Blot Washer System
Type Large Blot Tray
Material Polystyrene