missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IBA1 Polyclonal Antibody
GREENER_CHOICE

Numéro de catalogue. PIPA5143572
Modifier l'affichage
Click to view available options
Quantité:
100 μL
500 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
PIPA5143572 100 μL
PIPA5143538 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. PIPA5143572 Fournisseur Invitrogen™ Code fournisseur PA5143572

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Chicken Polyclonal Antibody

IBA1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Ionized calcium-binding adapter molecule 1 (IBA1), also known by its gene name AIF1, is a protein expressed predominantly by microglia in the brain and spinal cord. This protein belongs to the EF-hand calcium-binding protein family and plays a crucial role in microglial activation and migration in response to brain injury or neuroinflammation. IBA1's function is integral to microglial motility and phagocytic activity, facilitating the cellular response to pathogenic stimuli and promoting tissue homeostasis and repair in the central nervous system. IBA1 serves as a reliable marker for activated microglia in various neurological disorders, including Alzheimer's disease, Parkinson's disease, and multiple sclerosis, where increased expression correlates with disease progression and severity. The protein's structural features enable it to bind calcium ions, inducing conformational changes that activate signaling pathways essential for microglial function. Its expression is highly regulated by inflammatory cytokines, underpinning its role in neuroimmune responses. Due to its specific expression in microglia during pathological conditions, IBA1 is widely used in research as a marker to study microglial status and activity, and it remains a focal point for understanding microglial involvement in neurodegenerative diseases.

Spécifications

Antigène IBA1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugué Unconjugated
Formule PBS with 0.02% sodium azide
Gène AIF1
Numéro d’acquisition du gène O70200, P55008, P55009
Alias du gène AI607846; AIF1; AIF-1; Allograft inflammatory factor 1; balloon angioplasty responsive transcript; balloon angioplasty-responsive transcript 1; Bart1; BART-1; D17H6S50E; DADB-70P7.8; DASS-82G15.2; Em:AF129756.17; G1; Iba; Iba1; interferon gamma responsive transcript; ionized calcium binding adapter molecule 1; ionized calcium-binding adapter molecule 1; IRT1; IRT-1; microglia response factor; Mrf1; MRF-1; protein BART-1; protein G1; testis specific
Symboles de gène AIF1
Espèces hôtes Chicken
Immunogène Peptide identical to part of the C-terminal of human IBA1 coupled to KLH.
Quantité 100 μL
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 11629, 199, 29427
Espèces cibles Human, Mouse, Rat
Contenu et stockage 4°C
Type de produit Antibody
Forme Liquid
Isotype IgY
Afficher plus de résultats Afficher moins de résultats